Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-51 Mouse anti-Mouse, PE, Clone: BP-1, BD

Catalog No. BDB553735 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553735 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553735 Supplier BD Biosciences Supplier No. 553735
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The BP-1 antibody reacts with an epitope of the 6C3/BP-1 (Ly-51) glycoprotein cell-surface differentiation antigen, which was originally identified on pre-B cell lymphomas (spontaneous and chemical- or retrovirus- transformed, in vitro and in vivo). 6C3/BP-1 is a homodimer cell-surface glycoprotein with 140kDa subunits which has been identified to possess aminopeptidase A (APA) activity. The same antigen is expressed at high levels on bone marrow stromal cell lines which support in vitro B lymphopoieses, on thymic dendritic cells and cortical epithelial cells, and on a wide variety of mouse and rat tissues known to possess APA activity. Subsets of normal bone marrow pre-B and B lymphocytes express low levels of Ly-51, which is rapidly up-regulated on the pre-B cells in the presence of IL-7. A role for the 6C3/BP-1 molecule in the IL-7-driven proliferation of B cell precursors has been postulated. However, B-cell abnormalities were not detected in Ly-51-deficient mice. Mature B lymphocytes, thymocytes, peripheral T lymphocytes, erythroid cells, and myeloid cells (with the exception of thymic dendritic cells) do not express Ly-51.

Host Species: Mouse
Clone: BP-1
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Abelson murine leukemia virus-transformed BALB/c mouse pre-B cell line 18.81

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-51
Applications Flow Cytometry
Classification Monoclonal
Clone BP-1
Concentration 0.2mg/mL
Conjugate PE
Description 6C3/BP-1 Antigen
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Abelson murine leukemia virus-transformed BALB/c mouse pre-B cell line 18.81
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.