Learn More
Description
Host Species: Rat
Clone D7
Isotype IgG2a, κ
Species Reactivity Ms
Immunogen IL-2-dependent mouse T-cell line CTL-L
Flow Cytometry
Specifications
Specifications
| Antigen | Ly-6A/E |
| Classification | Monoclonal |
| Clone | D7 |
| Concentration | 0.2mg/mL |
| Host Species | Rat |
| Immunogen | IL-2-dependent mouse T-cell line CTL-L |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.