Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6A/E Rat anti-Mouse, Unlabeled, Clone: E13-161.7, BD

Catalog No. BDB553333 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553333 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553333 Supplier BD Biosciences Supplier No. 553333
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The E13-161.7 antibody reacts with Ly-6A.2 and Ly-6E.1, which are allelic members of the Ly-6 multigene family. Sca1 (Ly-6A/E), a phosphatidylinositol-anchored protein of about 18kDa, is expressed on the multipotent hematopoietic stem cell (HSC) in mice w

Host Species: Rat
Clone: E13-161.7
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: BALB/c mouse-derived “pre-T” cell hybridoma

Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6A/E
Applications Flow Cytometry
Classification Monoclonal
Clone E13-161.7
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Sca-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen BALB/c mouse-derived “pre-T” cell hybridoma
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.