Learn More
Ly-6C Rat anti-Mouse, Alexa Fluor 700, Clone: AL-21, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 561237
Description
The AL-21 monoclonal antibody specifically binds to a non-polymorphic determinant of Ly-6C, a 14-17 kDa GPI-linked cell-surface antigen found on some monocyte/macrophage populations, granulocytes, endothelial cells, plasma cells, and thymocyte, NK-cell, and T-subsets. Mice with the Ly-6.2 alloantigen (eg, AKR, C57BL, C57BR, C57L, C58, DBA/2, PL, SJL, SWR, 129) have subsets of CD8+ and CD4+ Ly-6C+ T cells, while Ly-6.1 strains (eg, A, BALB/c, CBA, C3H/He, DBA/1, NZB) have only CD8+ Ly-6C+ T cells. Upregulation of Ly-6C expression on CD8+ T cells by interferons α and β and poly (I:C) has been described, and Ly-6C is a memory marker on CD8+ T cells.

Specifications
Ly-6C | |
Monoclonal | |
0.2mg/mL | |
Ly6c; Lymphocyte antigen 6 complex, locus C; Lymphocyte antigen Ly-6C | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgM κ |
Flow Cytometry | |
AL-21 | |
Alexa Fluor 700 | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Not reported | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.