Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Ly-6G FITC, Clone: RB6-8C5,

Catalog No. MABF1481MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1481MI 25 μg
MABF1482MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF1481MI Supplier MilliporeSigma™ Supplier No. MABF1481
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects Ly-6G Clone: RB6-8C5 in Mouse samples, and it is validated for Flow Cytometry

The RB6-8C5 antibody binds to mouse Ly-6G, commonly known as Gr-1, a member of the Ly-6 superfamily of GPI-anchored cell surface proteins with roles in cell signaling and cell adhesion. Gr-1 is differentially expressed during development and maturation of cells in the myeloid lineage and is expression at varying stages and levels on monocytes, macrophages, granulocytes, and peripheral neutrophils. In the mouse, the RB6-8C5 antibody is typically used in combination with the macrophage labeling antibody M1/70 (Anti-CD11b) for phenotypic analysis of monocytes, macrophages and granulocytes.Note: The RB6-8C5 antibody has been reported to cross-react with Ly-6C on cells expressing this antigen (Fleming et al. 1993. J. Immunol. 151:2399-2408 and Sasmono et al. 2007. J. Leukoc. Biol. 82: 111-123) and has been cited in the literature for identification of Ly-6G/Ly-6C. Other reports suggest that this antibody is specific for Ly-6G, without cross-reactivity for Ly-6C (Nagendra S. and Schlueter AJ. 2003. Cytometry A, 58(2): 195-200.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6G
Applications Flow Cytometry
Classification Monoclonal
Clone RB6-8C5
Conjugate FITC
Formulation Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Gene Accession No. P35461
Gene Symbols Ly6g
Host Species Rat
Immunogen Proprietary immunogen
Purification Method Purified by size-exclusion chromatography
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation, Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.