Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly6C Rat anti-Mouse, Clone: 6C3, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABN668
Description
Detect Lymphocyte antigen 6C2 using this rat monoclonal antibody, Anti-Ly6C Antibody, clone 6C3 validated for use in Flow Cytometry and Dot Blot.
Specifications
Ly6C | |
Monoclonal | |
Unconjugated | |
NP_001092687 | |
Rat | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG2a κ |
Dot Blot, Flow Cytometry | |
6C3 | |
Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Ly6c2, Ly6c | |
Whole cells expressing mouse Ly6C. | |
100 μL | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction