Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

lymphocyte antigen 86, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016978 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-978 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-978 Supplier Abnova Corporation Supplier No. H00009450B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human LY86 protein.

Sequence: MKGFTATLFLWTLIFPSCSGGGGGKAWPTHVVCSDSGLEVLYQSCDPLQDFGFSVEKCSKQLKSNINIRFGIILREDIKELFLDLALMSQGSSVLNFSYPICEAALPKFSFCGRRKGEQIYYAGPVNNPEFTIPQGEYQVLLELYTEKRSTVACANATIMCS

Specifications

Antigen lymphocyte antigen 86
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human LY86 protein.
Formulation PBS with no preservative; pH 7.4
Gene LY86
Gene Accession No. NM_004271
Gene Alias MD-1/MMD-1/dJ80N2.1
Gene Symbols LY86
Host Species Mouse
Immunogen LY86 (NP_004262.1, 1 a.a. ∼ 162 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9450
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.