Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYAR Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB4344MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB4344MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB4344MI Supplier MilliporeSigma Supplier No. AB4344
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-LYAR Antibody (Ly1 reactive clone) detects level of LYAR (Ly1 antibody reactive clone) and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYAR (Ly1 reactive clone)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols Lyar; LYAR
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to mouse LYAR.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_079557
Target Species Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.