Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYAR, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017126 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-126 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-126 Supplier Abnova Corporation Supplier No. H00055646B02P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human LYAR protein.

Sequence: MVFFTCNACGESVKKIQVEKHVSVCRNCECLSCIDCGKDFWGDDYKNHVKCISEDQKYGGKGYEGKTHKGDIKQQAWIQKISELIKRPNVSPKVRELLEQISAFDNVPRKKAKFQNWMKNSLKVHNESILDQVWNIFSEASNSEPVNKEQDQRPLHPVANPHAEISTKVPASKVKDAVEQQGEVKKNKRERKEERQKKRKREKKELKLENHQENSRNQKPKKRKKGQEADLEAGGEEVPEANGSAGKRSKKKKQRKDSASEEEARVGAGKRKRRHSEVETDSKKKKMKLPEHPEGGEPEDDEAPAKGKFNWKGTIKAILKQAPDNEITIKKLRKKVLAQYYTVTDEHHRSEEELLVIFNKKISKNPTFKLLKDKVKLVK

Specifications

Antigen LYAR
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human LYAR protein.
Formulation PBS with no preservative; pH 7.4
Gene LYAR
Gene Accession No. NM_017816
Gene Alias FLJ20425/ZLYAR
Gene Symbols LYAR
Host Species Mouse
Immunogen LYAR (NP_060286, 1 a.a. ∼ 379 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55646
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.