Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lyn Mouse, Unlabeled, Clone: 42, BD

Catalog No. BDB610003 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610003 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610003 Supplier BD Biosciences Supplier No. 610003

Mouse Monoclonal Antibody

Lyn is a 56 kDa cytosolic tyrosine kinase that belongs to the Src family of kinases. In addition to Src, this family includes lck, Fgr, Hck, Blk, Yes, fyn, and Yrk kinases. The lyn gene was isolated from a human placenta cDNA library and later found in a variety of cell types. Similar to other members of the Src family of tyrosine kinases, Lyn has been implicated in the regulation of cell signaling mechanisms. Lyn's function has primarily been studied in hematopoietic cells where it is associated with the high-affinity IgE receptor in basophils. It also associates with p120 Ras-GAP in thrombin-stimulated platelets. This association corresponds to an increase in Lyn kinase activity. Lyn is often observed as a doublet of about 56 kDa and 53 kDa. These forms may represent alternative splicing products of lyn mRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lyn
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 42
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Lyn aa. 1-138
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.