Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lyn Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 06207MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-207-MI 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-207-MI Supplier MilliporeSigma Supplier No. 06207
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Lyn Antibody detects level of Lyn and has been published and validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lyn
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation whole antiserum containing 0.05% sodium azide
Gene Symbols LYN; FLJ26625; JTK8
Host Species Rabbit
Immunogen peptide (NKQQRPVPESQL-LPGQRFQT) corresponding to residues 39-58 of human Lyn
Purification Method Antiserum
Quantity 250 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002350
Target Species Human
Content And Storage −20°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.