Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYST, Mouse, Clone: 3H1, Abnova™

Catalog No. 89021081 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-081 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-081 Supplier Abnova Corporation Supplier No. H00001130M07
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant LYST.

This gene is thought to encode a protein that regulates intracellular protein trafficking to and from the lysosome. Mutations in this gene are associated with Chediak-Higashi syndrome, a rare lysosomal storage disorder. Alternatively spliced transcript variants have been described but their full-length nature has not been determined. [provided by RefSeq

Sequence: MSTDSNSLAREFLTDVNRLCNAVVQRVEAREEEEEETHMATLGQYLVHGRGFLLLTKLNSIIDQALTCREELLTLLLSLLPLVWKIPVQEEKATDFNLPL

Specifications

Antigen LYST
Applications ELISA
Classification Monoclonal
Clone 3H1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant LYST.
Formulation PBS with no preservative; pH 7.4
Gene LYST
Gene Accession No. NM_000081
Gene Alias CHS/CHS1
Gene Symbols LYST
Host Species Mouse
Immunogen LYST (NP_000072.2, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1130
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.