Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYVE1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB2988MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2988MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2988MI Supplier MilliporeSigma Supplier No. AB2988
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-LYVE1 Antibody is an antibody against LYVE1 for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYVE1
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Symbols LYVE1; CRSBP-1; CRSBP1; HARl; LYVE-1; XLKD1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to mouse LYVE1.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_444477
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.