Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-LYZ, Polyclonal,
Supplier: MilliporeSigma™ ABF224
Description

Specifications
LYZ | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide and 50% glycerol. | |
Rabbit | |
Affinity Purified | |
Primary | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
Western Blot | |
Unconjugated | |
LYZ; LZM | |
GST-tagged fusion protein corresponding to human LYZ. | |
150 μL | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction