Learn More
LYZL2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human LYZL2 peptide using ARM Technology.
Supplier: Abnova Corporation H00119180K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Lysozymes (see LYZ; MIM 153450), especially C-type lysozymes, are well-recognized bacteriolytic factors widely distributed in the animal kingdom and play a mainly protective role in host defense. LYZL2 is a member of a family of lysozyme-like genes (Zhang et al., 2005 [PubMed 16014814]).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
LYZL2 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
LYZL2 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
1mg | |
lysozyme-like 2 | |
LYZL2 | |
LYZL2 | |
A synthetic peptide of human LYZL2 | |
RUO | |
119180 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.