Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LZTS1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535130
Description
This gene encodes a tumor suppressor protein that is ubiquitously expressed in normal tissues. In uveal melanomas, expression of this protein is silenced in rapidly metastasizing and metastatic tumor cells but has normal expression in slowly metastasizing or nonmetastasizing tumor cells. This protein may have a role in cell-cycle control by interacting with the Cdk1/cyclinB1 complex. This gene is located on chromosomal region 8p22. Loss of heterozygosity (LOH) in the 8p arm is a common characteristic of many types of cancer.
Specifications
| LZTS1 | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| Q9Y250, P60853 | |
| LZTS1 | |
| KLH conjugated synthetic peptide between 77-105 amino acids from the N-terminal region of human LZTS1 | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| LZTS1 | |
| LZTS1; FEZ1; Leucine zipper putative tumor suppressor 1; F37/esophageal cancer-related gene-coding leucine-zipper motif; Fez1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11178, 211134 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction