Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M-Cadherin Mouse anti-Mouse, Clone: 12G4, MilliporeSigma™

Catalog No. 05852MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-852-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-852-MI Supplier MilliporeSigma Supplier No. 05852
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-M-Cadherin Antibody, clone 12G4 is an antibody against M-Cadherin for use in ELISA, western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen M-Cadherin
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 12G4
Conjugate Unconjugated
Gene Accession No. NM_007662
Gene Symbols CDH15, CDH3, myotubule-cadherin, cadherin-14, Muscle-cadherin, CDHM, cadherin-3, muscle-cadherin, Cadherin-14, M-cadherin, CDH14, MCAD
Host Species Mouse
Immunogen Recombinant extracellular domain of mouse M-Cadherin
Purification Method Thiophilic adsorption and size exclusion
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage −20°C for up to 12 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.