Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M-Cadherin Mouse, Unlabeled, Clone: 5, BD

Catalog No. BDB611101 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611101 150 μg
BDB611100 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611101 Supplier BD Biosciences Supplier No. 611101
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cadherins are a family of transmembrane glycoproteins involved in the Ca2+;-dependent cell-cell adhesion that occurs in many tissues. Members of this family include P-Cadherin, E-Cadherin (uvomorulin), N-Cadherin, R-Cadherin, Cadherin-5, L-CAM, and EP-Cadherin. These proteins are similar in their domain structure (45-74% amino acid conservation), Ca2+; and protease sensitivity, and molecular weight. However, cadherins have distinct tissue expression patterns and immunological reactivities. M (muscle)-Cadherin, another member of the Cadherin family, was discovered in myogenic mouse cells where it is present at low levels in myoblasts. It is expressed in prenatal and adult skeletal muscle and plays a role in skeletal muscle cell differentiation, particularly the fusion of myoblasts into myotubes. It is upregulated
upon induction of myotube formation. M-Cadherin also forms complexes with the catenins in skeletal muscle cells, which then interact with the cytoskeleton. Therefore, it is thought that the M-Cadherin-cytoskeleton interaction may play a role in aligning myoblasts during fusion.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen M-Cadherin
Applications Western Blot
Classification Monoclonal
Clone 5
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse M-Cadherin aa. 253-366
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.