Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
M-CSFR/CD115 Mouse anti-Human, Fluorescein, Clone: 61708, R&D Systems™
Mouse Monoclonal Antibody
Specifications
| Antigen | M-CSFR/CD115 |
|---|---|
| Clone | 61708 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
FAB329F025
|
R&D Systems
FAB329F025 |
25 Tests | N/A | N/A | N/A | ||||
|
FAB329F100
|
R&D Systems
FAB329F100 |
100 Tests | N/A | N/A | N/A | ||||
Description
M-CSF R/CD115 Monoclonal antibody specifically detects M-CSF R/CD115 in Human samples. It is validated for Flow Cytometry.Specifications
| M-CSFR/CD115 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF1R, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, MCSFR, M-CSFR, Proto-oncogene c-Fms | |
| CSF1R | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human M-CSF R in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) GM-CSF R alpha or rhGM-CSF R beta . |
| 61708 | |
| Flow Cytometry | |
| Fluorescein | |
| Mouse | |
| Human | |
| P07333.2 | |
| 1436 | |
| Mouse myeloma cell line NS0-derived recombinant human M-CSF R Ile20-Glu512 (Pro54Ala) Accession # P07333.2 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title