Learn More
IL-23 p19 Rat anti-Mouse, Clone: G23-8, eBioscience
Rat Monoclonal Antibody
Manufacturer: Invitrogen 14723285
Description
Description: The G23-8 antibody reacts with the p19 subunit of mouse IL-23. The G23-8 antibody was generated from immunization with authentic, insect cell-expressed, recombit mouse IL-23 heterodimer. The G23-8 antibody can specifically neutralize IL-23 bioactivity with no effect on IL-12 p70 bioactivity. The use of a p19-specific capture antibody and a p40-specific detection antibody yields an IL-23 ELISA which is exquisitely specific for mouse IL-23. IL-12 p40 homodimer and IL-12 p70 were each run in the assay at 500 ng/mL with no interference or cross-reactivity observed. A panel of 20 unrelated cytokines was run in the IL-23 ELISA at 100 ng/mL with no cross reactivity observed; all values were at the limit of detection of the assay. For measurement of total p40 protein levels, the Mouse IL-12/23 Total p40 ELISA Ready-SET-Go! Is available (88-7120). IL-23 is a heterodimeric cytokine composed of the p40 subunit of IL-12 disulfide-linked with a protein p19. p19, like p35 of IL-12, is biologically inactive by itself. IL-23 interacts with IL-12Rbeta1 and an additional, novel beta2-like receptor subunit with STAT4 binding domain, termed IL-23R. IL-23 is secreted by activated mouse and human dendritic cells. Biological activities of mouse IL-23 are distinct from those of mouse IL-12. Mouse IL-23 was found not to induce significant amounts of IFN-g.
Mouse IL-23 does induce strong proliferation of memory T cells (but not naive T cells), whereas IL-12 has no effect on memory cells. Additionally, mouse IL-23 (but not IL-12) can activate mouse memory T cells to produce the proinflammatory cytokine IL-17. Human IL-23 has biological properties which are less distinct from human IL-12; human IL-23 induces proliferation of memory T cells and induces moderate levels of IFN-g production by naive and memory T cells, as compared to IL-12. Applications Reported: The G23-8 antibody has been reported for use as the capture antibody in mouse IL-23 ELISA, for Western blotting, and for neutralization of mouse IL-23 bioactivity. Applications Tested: The G23-8 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse IL-23 (p19p40) protein levels in combination with the biotinylated (p40-specific) C17.8 antibody (13-7123) for detection and recombit mouse IL-23 (14-8231) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1-4 µg/mL. A standard curve consisting of doubling dilutions of the recombit standard over the range of 4000 pg/mL - 30 pg/mL should be included in each ELISA plate. Important Note: TMB, rather than ABTS, should be used as a substrate to achieve this sensitivity level. For specific neutralization of mouse IL-23 protein activity (with no effect on IL-12 p70), the functional grade purified G23-8 antibody is recommended (cat No.16-7232). Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2 µm post-manufacturing filtered. The p19 shares sequence similarity with IL-6 subfamily members and is distantly related to the p35 subunit of IL-12. It shows no biological activity by itself; instead, it combines with the p40 subunit of IL-12 to form a biologically active, composite cytokine, IL-23. IL-23 shares some in vivo functions with IL-12, including the activation of the transcription factor Stat4. Similar to IL-12, human IL-23 stimulates IFN-g production and proliferation of PHA blast T cells, as well as in CD45 RO (memory) T cells. Ubiquitous transgenic expression of the IL-23 subunit p19 induces multiorgan inflammation, runting, infertility and premature death.Specifications
IL-23 p19 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9EQ14 | |
IL-23, IL-23A, IL23P19, MGC79388, P19, SGRF | |
Rat | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Mouse |
ELISA, Western Blot | |
G23-8 | |
Unconjugated | |
Il23a | |
Liquid | |
Il23a | |
IgG1, kappa | |
500 μg | |
4° C | |
Primary | |
83430 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.