Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mA20 Rat anti-Mouse, Clone: 37, MilliporeSigma™

Catalog No. MABF220MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF220MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF220MI Supplier MilliporeSigma Supplier No. MABF220
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect mA20/TNFAIP3 using this Anti-mA20 Antibody/TNFAIP3, clone 37 validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mA20/TNFAIP3
Applications Flow Cytometry
Classification Monoclonal
Clone 37
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Purified rat monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_033423
Gene Symbols Tnfaip3, Tnfip3
Host Species Rat
Immunogen Recombinant protein corresponding to Mouse mA20/TNFAIP3.
Purification Method Purified by ion-exchange chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.