Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

macro-H2A1.2 Mouse anti-Human, Clone: 14G7, MilliporeSigma™

Catalog No. MABE61MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE61MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE61MI Supplier MilliporeSigma Supplier No. MABE61
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-macro-H2A1.2 Antibody, clone 14G7 is a Mouse Monoclonal Antibody for detection of macro-H2A1.2 also known as H2A histone family member Y or Histone macroH2A1 and has been validated in western blot, immunocytochemistry and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen macro-H2A1.2
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 14G7
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bλ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001035248
Gene Symbols H2A.y, H2A/y, H2AF12M, H2AFJ, MACROH2A1, MACROH2A1.1, mH2A1, macroH2A1.2, macro-H2A1.2
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human macro-H2A1.2.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.