Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Macrophage Subset Mouse anti-Rat, PE, Clone: HIS36, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 554901
Description
The HIS36 antibody reacts with an ED2-like antigen, which is found on tissue macrophages and thioglycollate-elicited peritoneal exudate cells, but not on monocytes.

Specifications
Macrophage Subset | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Not reported | |
0.2 mg | |
Primary | |
Rat | |
IgG2a κ |
Flow Cytometry | |
HIS36 | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
PE Mouse Anti-Rat Macrophage Subset, PE Mouse IgG2a, κ Isotype Control (0.1mg) | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction