Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Macrophages/Granulocytes Mouse anti-Human, Mouse, Rat, Clone: OX-41, MilliporeSigma™

Catalog No. MAB1407PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1407PMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1407PMI Supplier MilliporeSigma Supplier No. MAB1407P
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Macrophages/Granulocytes Antibody, clone OX-41 is validated for use in flow cytometry, immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin), western blotting for the detection of Macrophages/Granulocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Macrophages/Granulocytes
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OX-41
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2a in buffer containing 0.09% sodium azide.
Gene Accession No. NP_001035111
Gene Symbols CD172a, SIRPA, SIRPalpha2, BIT, SHPS1, MFR, SIRP, p84, SHPS-1, Sirp-alpha-1, CD172A, Sirp-alpha-2, SIRP-ALPHA-1, PTPNS1, SIRPalpha, P84, Bit, MYD1, MYD-1
Host Species Mouse
Immunogen Rat peritoneal macrophages.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.