Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Macrophages/Monocytes Mouse anti-Rat, Clone: ED-1, MilliporeSigma™

Catalog No. MAB1435MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1435MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1435MI Supplier MilliporeSigma Supplier No. MAB1435
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Macrophages/Monocytes Antibody, clone ED-1 is validated for use in Flow Cytometry (FC), Immunohistochemistry (immunohistochemistry), Immunoprecipitation (IP), Radioimmunoassay (RIA), and Western Blotting (WB) for the detection of CD68.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Macrophages/Monocytes
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone ED-1
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Host Species Mouse
Immunogen Rat spleen cells
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.