Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mad2 Mouse anti-Human, Clone: AS55-A12, MilliporeSigma™

Catalog No. MABE866MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE866MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE866MI Supplier MilliporeSigma Supplier No. MABE866
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Mad2 Antibody, clone AS55-A12 is a highly specific mouse monoclonal antibody, that targets MAD2 and has been tested in western blotting and Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mad2
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone AS55-A12
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2b in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_002349
Gene Symbols MAD2L1, MAD2
Host Species Mouse
Immunogen His-tagged recombinant protein corresponding to human Mad2.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.