Learn More
MAdCAM-1 Rat anti-Mouse, Unlabeled, Clone: MECA-367, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 550556
Description
The MECA-367 antibody reacts with mucosal vascular addressin MAdCAM-1. In the fetus and neonate, MAdCAM-1 is the predominant vascular addressin on the high endothelial venules (HEV) of peripheral lymph nodes. In adult mice, MAdCAM-1 is preferentially expr
Specifications
| MAdCAM-1 | |
| Monoclonal | |
| 31.25μg/mL | |
| Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide. | |
| Mouse BALB/c mesenteric and peripheral lymph nodes | |
| 1 mL | |
| Cell Biology | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| MECA-367 | |
| Unconjugated | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.