Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MADD Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. PIPA129177
Change view
Click to view available options
Quantity:
500 μL
Catalog No. Quantity
PIPA129177 500 μL
1 options

Catalog No. PIPA129177

Supplier: Thermo Scientific PA129177

Rabbit Polyclonal Antibody

PA1-29178 detects MMP1 from human samples. PA1-29178 has been successfully used in immunohistochemistry (paraffin tissue) and Western blot applications. The PA1-29178 immunogen is a synthetic peptide from the middle region of the human MMP-1. Positive

Tumor necrosis factor receptor 1 (TNFR1) contains a death receptor domain that mediates interactions involved in downstream signaling of TNF. A novel protein, MADD (mitogen activated protein (MAP)-kinase activating death domain protein) has recently been
TRUSTED_SUSTAINABILITY

Specifications

Antigen MADD
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene MADD
Gene Accession No. Q8WXG6
Gene Alias MAP-Kinase Activating Death Domain, DENN, Differentially Expressed in Neoplasmic vs. Normal Cells
Gene Symbols MADD
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the second half of human MADD protein.
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8567
Target Species Human
Content And Storage 4° C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.