Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-MafK/Nfe2u, Polyclonal

Catalog No. ABE1928MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE1928MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1928MI Supplier MilliporeSigma™ Supplier No. ABE1928
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects MafK/Nfe2u Clone: in Human, Mouse, Rat samples, and it is validated for ChIP, Chromatin Immunoprecipitation, Western Blotting

Transcription factor MafK (UniProt Q61827; also known as Erythroid transcription factor NF-E2 p18 subunit, Nuclear factor erythroid derived 2 ubiquitous, p18 NF-E2) is encoded by the Mafk (also known as Nfe2u, AW061068) gene (Gene ID 17135) in murine species. MafK, MafG and MafF consistute a family of transcription factors that dimerize with the CNC (cap' n' collar) family regulatory protein Nrf2 (NF-E2 p45-related factor 2) via their bZip domain. The Nrf2/small Maf heterodimer regulates downstream genes, including those of phase II detoxifying enzymes, by targeting antioxidant responsive element (ARE) and Maf recognition element (MARE).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MafK/Nfe2u
Applications ChIP Assay, Western Blot, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. Q61827
Gene Symbols Mafk, Nfe2u
Host Species Rabbit
Immunogen Maltose binding protein-tagged recombinant protein corresponding to mouse MafK/Nfe2u.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_034887
Target Species Human, Mouse, Rat
Content And Storage −20°C for one year
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.