Learn More
myelin associated glycoprotein, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MAG protein.
Supplier: Abnova Corporation H00004099B01
Description
The protein encoded by this gene is a type I membrane protein and member of the immunoglobulin superfamily. It is thought to be involved in the process of myelination. It is a lectin that binds to sialylated glycoconjugates and mediates certain myelin-neuron cell-cell interactions. Two alternatively spliced transcripts encoding different isoforms have been described for this gene. [provided by RefSeq
Sequence: MIFLTALPLFWIMISASRGGHWGAWMPSSISAFEGTCVSIPCRFDFPDELRPAVVHGVWYFNSPYPKNYPPVVFKSRTQVVHESFQGRSRLLGDLGLRNCTLLLSNVSPELGGKYYFRGDLGGYNQYTFSEHSVLDIVNTPNIVVPPEVVAGTEVEVSCMVPDNCPELRPELSWLGHEGLGEPAVLGRLREDEGTWVQVSLLHFVPTREANGHRLGCQASFPNTTLQFEGYASMDVKYPPVIVEMNSSVEAIEGSHVSLLCGADSNPPPLLTWMRDGTVLREAVAESLLLELEEVTPAEDGVYACLAENAYGQDNRTVGLSVMYAPWKPTVNGTMVAVEGETVSILCSTQSNPDPILTIFKEKQILSTVIYESELQLELPAVSPEDDGEYWCVAENQYGQRATAFNLSVEFAPVLLLESHCAAARDTVQCLCVVKSNPEPSVAFELPSRNVTVNESEREFVYSERSGLVLTSILTLRGQAQAPPRVICTARNLYGAKSLELPFQGAHRLMWAKIGPVGAVVAFAILIAIVCYITQTRRKKNVTESPSFSAGDNPPVLFSSDFRISGAPEKYESERRLGSERRLLGLRGEPPELDLSYSHSDLGKRPTKDSYTLTEELAEYAEIRVKSpecifications
myelin associated glycoprotein | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human MAG protein. | |
MAG | |
GMA/S-MAG/SIGLEC-4A/SIGLEC4A | |
Mouse | |
50 μL | |
Cell Adhesion | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_002361.2 | |
MAG | |
MAG (NP_002352.1, 1 a.a. ∼ 626 a.a) full-length human protein. | |
RUO | |
Yes | |
4099 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.