Learn More
MAGEA3, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human MAGEA3 peptide using ARM Technology.
Supplier: Abnova Corporation H00004102K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
| MAGEA3 | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| MAGEA3 | |
| MAGEA3 | |
| A synthetic peptide of human MAGEA3 | |
| RUO | |
| 4102 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| ELISA, Western Blot | |
| 1mg | |
| melanoma antigen family A, 3 | |
| MAGEA3 | |
| HIP8, HYPD, MAGE3, MAGEA6, MGC14613 | |
| Rabbit | |
| 1 mg x up to 3 Abs | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.