Learn More
melanoma antigen family B, 6, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MAGEB6 protein.
Supplier: Abnova Corporation H00158809B01
Description
This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. This gene is expressed in testis, and in a significant fraction of tumors of various histological types. The MAGEB genes are clustered on chromosome Xp22-p21. [provided by RefSeq
Sequence: MPRGHKSKLRTCEKRQETNGQPQGLTGPQATAEKQEESHSSSSSSRACLGDCRRSSDASIPQESQGVSPTGSPDAVVSYSKSDVAANGQDEKSPSTSRDASVPQESQGASPTGSPDAGVSGSKYDVAANGQDEKSPSTSHDVSVPQESQGASPTGSPDAGVSGSKYDVAAEGEDEESVSASQKAIIFKRLSKDAVKKKACTLAQFLQKKFEKKESILKADMLKCVRREYKPYFPQILNRTSQHLVVAFGVELKEMDSSGESYTLVSKLGLPSEGILSGDNALPKSGLLMSLLVVIFMNGNCATEEEVWEFLGLLGIYDGILHSIYGDARKIITEDLVQDKYVVYRQVCNSDPPCYEFLWGPRAYAETTKMRVLRVLADSSNTSPGLYPHLYEDALIDEVERALRLRASpecifications
melanoma antigen family B, 6 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human MAGEB6 protein. | |
MAGEB6 | |
FLJ40242/MAGE-B6/MAGEB6A | |
Mouse | |
50 μL | |
Yes | |
158809 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Western Blot | |
Unconjugated | |
No additive | |
NM_173523.2 | |
MAGEB6 | |
MAGEB6 (NP_775794.2, 1 a.a. ∼ 407 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.