Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

melanoma antigen family D, 1 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89006791 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-006-791 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-006-791 Supplier Abnova Corporation Supplier No. H00009500B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human MAGED1 protein.

This gene is a member of the melanoma antigen gene (MAGE) family. Most of the genes of this family encode tumor specific antigens that are not expressed in normal adult tissues except testis. Although the protein encoded by this gene shares strong homology with members of the MAGE family, it is expressed in almost all normal adult tissues. This gene has been demonstrated to be involved in the p75 neurotrophin receptor mediated programmed cell death pathway. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq

Sequence: MAQKMDCGAGLLGFQAEASVEDSALLMQTLMEAIQISEAPPTNQATAAASPQSSQPPTANEMADIQVSAAAARPKSAFKVQNATTKGPNGVYDFSQAHNAKDVPNTQPKAAFKSQNATPKGPNAAYDFSQAATTGELAANKSEMAFKAQNATTKVGPNATYNFSQSLNANDLANSRPKTPFKAWNDTTKAPTADTQTQNVNQAKMATSQADIETDPGISEPDGATAQTSADGSQAQNLESRTIIRGKRTRKINNLNVEENSSGDQRRAPLAAGTWRSAPVPVTTQNPPGAPPNVLWQTPLAWQNPSGWQNQTARQTPPARQSPPARQTPPAWQNPVAWQNPVIWPNPVIWQNPVIWPNPIVWPGPVVWPNPLAWQNPPGWQTPPGWQTPPGWQGPPDWQGPPDWPLPPDWPLPPDWPLPTDWPLPPDWIPADWPIPPDWQNLRPSPNLRPSPNSRASQNPGAAQPRDVALLQERANKLVKYLMLKDYTKVPIKRSEMLRDIIREYTDVYPEIIERACFVLEKKFGIQLKEIDKEEHLYILISTPESLAGILGTTKDTPKLGLLLVILGVIFMNGNRASEAVLWEALRKMGLRPGVRHPLLGDLRKLLTYEFVKQKYLDYRRVPNSNPPEYEFLWGLRSYHETSKMKVLRFIAEVQKRDPRDWTAQFMEAADEALDALDAAAAEAEARAEARTRMGIGDEAVSGPWSWDDIEFELLTWDEEGDFGDPWSRIPFTFWARYHQNARSRFPQTFAGPIIGPGGTASANFAANFGAIGFFWVE

Specifications

Antigen melanoma antigen family D, 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human MAGED1 protein.
Formulation No additive
Gene MAGED1
Gene Accession No. NM_001005332.1
Gene Alias DLXIN-1/NRAGE
Gene Symbols MAGED1
Host Species Mouse
Immunogen MAGED1 (NP_001005332.1, 1 a.a. ∼ 778 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 9500
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.