Learn More
Description
Includes
Up to three rabbit IgG clones of 1mg each will be delivered to customer.
This gene is a member of the MAGED gene family. It is expressed only in brain and ovary among normal tissues, and two transcript variants of this gene are specifically expressed in glioma cells among cancer cells. This gene and the other MAGED genes are clustered on chromosome Xp11. Multiple alternatively spliced transcript variants have been found for this gene, however, the full-length nature of some variants has not been defined. [provided by RefSeq
Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
Specifications
| Antigen | MAGED4B |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Concentration | 1mg |
| Conjugate | Unconjugated |
| Description | melanoma antigen family D, 4B |
| Formulation | In 1X PBS, pH 7.4 |
| Gene | MAGED4B |
| Gene Accession No. | MAGED4B |
| Gene Alias | MGC3210, MGC88639 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.