Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGOH Mouse anti-Human, Clone: 21B12, MilliporeSigma™

Catalog No. 051538MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-153-8MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-153-8MI Supplier MilliporeSigma Supplier No. 051538

Mouse Monoclonal Antibody

Use Anti-MAGOH Antibody, clone 21B12 (Mouse Monoclonal Antibody) validated in western blot, immunoprecipitation, immunocytochemistry to detect MAGOH also known as mago-nashi (Drosophila) homolog proliferation-associated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGOH
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 21B12
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgGκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_002361.1
Gene Symbols MAGOH, MAGOHA
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human MAGOH.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.