Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

male germ cell-associated kinase, Mouse, Clone: 3E5, Abnova™

Catalog No. 89000792 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-792 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-792 Supplier Abnova Corporation Supplier No. H00004117M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant MAK.

The product of this gene is a serine/threonine protein kinase related to kinases involved in cell cycle regulation. It is expressed almost exclusively in the testis, primarily in germ cells. Studies of the mouse and rat homologs have localized the kinase to the chromosomes during meiosis in spermatogenesis, specifically to the synaptonemal complex that exists while homologous chromosomes are paired. There is, however, a study of the mouse homolog that has identified high levels of expression in developing sensory epithelia so its function may be more generalized. [provided by RefSeq

Sequence: PPKQQSQEKPPQTLFPSIVKNMPTKPNGTLSHKSGRRRWGQTIFKSGDSWEELEDYDFGASHSKKPSMGVFKEKRKKDSPFRQQVKMAVISLSAHQFPTL

Specifications

Antigen male germ cell-associated kinase
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3E5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant MAK.
Formulation PBS with no preservative; pH 7.4
Gene MAK
Gene Accession No. BC039825
Gene Alias dJ417M14.2
Gene Symbols MAK
Host Species Mouse
Immunogen MAK (AAH39825, 358 a.a. ∼ 457 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 4117
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.