Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maltose Binding Protein Mouse anti-Bacteria, Clone: 1H4, MilliporeSigma™

Catalog No. MAB3108MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3108MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3108MI Supplier MilliporeSigma Supplier No. MAB3108
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Maltose Binding Protein Antibody, clone 1H4 detects level of Maltose Binding Protein and has been published and validated for use in ELISA, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Maltose Binding Protein
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1H4
Conjugate Unconjugated
Formulation Liquid, does not contain any preservative.
Host Species Mouse
Immunogen MBP fusion protein
Quantity 100 μL
Primary or Secondary Primary
Target Species Bacteria
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.