Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Mannose 6-Phosphate Receptor Clone: 2G11,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ 444105100UG
Description
Specifically detects Mannose 6-Phosphate Receptor Clone: 2G11 in Bovine, Human, Monkey samples, and it is validated for Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Lysosomal enzymes containing one or two mannose 6-phosphate (man6P) moieties are moved about in the cell by two distinct but interconnected cycles by means of ∽300kDa cation-independent MPRs. The biosynthetic cycle refers to the MPR transport of newly synthesized lysosomal enzymes from the trans Golgi network to late endosomes or early lysosomes. The endocytic cycle transports extracellular lysosomal enzymes from the plasma membrane via clathrin-coated vesicles to early endosomes. The entire pool of MPRs cycles between these cellular compartments every 3 hours. The steady state distribution of MPRs is predominantly late endosomes, fewer in the trans Golgi network, and ∽10% at cell surface. In addition to its man6P binding activity, MPR contains a separate binding site for the type II insulin-like growth factor and is capable of binding both man6P and IGF-II simultaneously. An ∽240kDa soluble, truncated form of the extracellular domain of MPR has been detected circulating in serum that is capable of binding both ligands.
Specifications
Mannose 6-Phosphate Receptor | |
Monoclonal | |
Unconjugated | |
Mouse | |
100 μg | |
Primary | |
Bovine, Human, Monkey | |
Purified |
Immunofluorescence, Immunohistochemistry, Immunoprecipitation | |
2G11 | |
In PBS. | |
native bovine cation independent (CI)-mannose 6-phosphate receptor | |
RUO | |
Recognizes the ∽300kDa mannose 6-phosphate receptor protein. | |
−20°C | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction