Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mannosidase II Rabbit anti-Human, Monkey, Polyclonal, MilliporeSigma™

Catalog No. AB3712MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3712MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3712MI Supplier MilliporeSigma Supplier No. AB3712

Rabbit Polyclonal Antibody

Detect the Mannosidase II protein using this Anti-Mannosidase II, For Golgi Apparatus identification validated for use in immunocytochemistry and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mannosidase II
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum. Liquid with 0.01% azide.
Gene Symbols MAN2A1; GOLIM7; MANII; MANA2; EC 3.2.1.114
Host Species Rabbit
Immunogen Purified peptide from human mannosidase II.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002372.2
Target Species Human, Monkey
Content And Storage 2-8°C for up to 2 months, −70°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.