Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP Kinase 2/Erk2 Mouse anti-Avian, Human, Mouse, Rat, Clone: 1B3B9, MilliporeSigma™

Catalog No. 05157MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-157-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-157-MI Supplier MilliporeSigma Supplier No. 05157

Mouse Monoclonal Antibody

Anti-MAP Kinase 2/Erk2 Antibody, clone 1B3B9 is an antibody against MAP Kinase 2/Erk2 for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP Kinase 2/Erk2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1B3B9
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2a in buffer containing PBS with 0.05% sodium azide. Store frozen at −20°C, avoid freeze thaw cycles.
Gene Accession No. NM_002745
Gene Symbols MAPK1, p38, ERK, P42MAPK, ERK2, p40, PRKM1, p41mapk, PRKM2, MAPK2, p42-MAPK, p41, ERK-2, ERT1
Host Species Mouse
Immunogen HPLC purified murine recombinant MAP Kinase. Clone 1B3B9
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Avian, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.