Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP1 Mouse anti-Human, Mouse, Rat, Clone: HM-1, MilliporeSigma™

Catalog No. MAB362MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB362MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB362MI Supplier MilliporeSigma Supplier No. MAB362
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect MAP1 using this Anti-MAP1 Antibody, clone HM-1 validated for use in western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP1
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone HM-1
Conjugate Unconjugated
Formulation Ascites fluid. Liquid containing 15mM sodium azide.
Gene Accession No. NM_002373.5
Gene Symbols MAP1A, FLJ77111, MTAP1A, MAP1L
Host Species Mouse
Immunogen Rat brain microtubule-associated protein.
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.