Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP1 Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABC334
Description
Detect Modulator of apoptosis 1 using this rabbit polyclonal antibody, Anti-MAP1 Antibody validated for use in western blotting.
Specifications
MAP1 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
2-8°C for 12 months |
Western Blot | |
Unconjugated | |
MOAP1; PNMA4 | |
KLH-conjugated linear peptide corresponding to the intermediate domain of human MAP1. | |
100 μg | |
Human, Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction