Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP1B Mouse, Unlabeled, Clone: 6, BD

Catalog No. BDB612679 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612679 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612679 Supplier BD Biosciences Supplier No. 612679

Mouse Monoclonal Antibody

Microtubule-associated proteins (MAPs) play a crucial role in the development and structure of nerve cells. These proteins are important for the assembly and stability of microtubles during neurite outgrowth and for the morphology of neuronal processes, such as dendrites. MAP1B encodes for a single precursor protein that is cleaved to produce a single heavy chain of 280kDa and a light chain of 32kDa which become noncovalently associated. Expression levels of MAP1B are reportedly highest in the brain and are modulated during development as reported with gene silencing experiments that implicate MAP1B's role during neuronal differentiation. In addition, MAP1B can be selectively phosphorylated depending on the cell type, subcellular localization and developmental phase. In neurons, MAP1B is expressed as a glycoprotein at the plasma membrane, where it can interact with proteins participating in axonal guiding. MAP1B has been reported to be observable to migrate in a range between 320-340kDa.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP1B
Applications Western Blot
Classification Monoclonal
Clone 6
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse MAP1B aa. 1745-1858
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.