Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP1LC3 C Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB15414
Description
This Anti-MAP1LC3 C Antibody is validated for use in western blot, immunocytochemistry, immunohistochemistry for the detection of MAP1LC3 C.
Specifications
MAP1LC3 C | |
Polyclonal | |
Lyophilized. Reconstitute with 50 uL of sterile distilled water. Centrifuge to remove any insoluble material. Contains no preservative. | |
Rabbit | |
Serum | |
Primary | |
Human, Rat | |
Serum |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
MAP1LC3C | |
Synthetic peptide corresponding to a region within the N-terminus of human MAP1LC3 C protein. | |
50 μL | |
NM_001004343.1 | |
Lyophilized: −20-−70°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction