Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2 Chicken anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB15452MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15452MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15452MI Supplier MilliporeSigma Supplier No. AB15452
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Anti-MAP2 Antibody detects level of MAP2 and has been published and validated for use in immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP2
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified chicken immunoglobulin. Two different antibodies were combined to make this product. Liquid in PBS, pH 7.2 with 0.02% sodium azide.
Gene Symbols MAP2; MAP2B; MAP-2; MAP2C; DKFZp686I2148; MAP2A
Host Species Chicken
Immunogen Synthetic peptides corresponding to different regions of the human/mouse MAP2 protein.
Purification Method Affinity Purified
Quantity 500 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001039538.1; NM_002374.3; NM_031845.2; NM_031847.2
Target Species Mouse
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.