Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2 Mouse anti-Bovine, Chicken, Human, Mouse, Rat, AlexaFluor™488, Clone: AP20, MilliporeSigma™

Catalog No. MAB3418XMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3418XMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3418XMI Supplier MilliporeSigma Supplier No. MAB3418X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-MAP2 Antibody, Alexa Fluor 488 conjugated is validated for use in immunohistochemistry for the detection of MAP2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP2
Applications Immunohistochemistry
Classification Monoclonal
Clone AP20
Conjugate Alexa Fluor 488
Formulation Purified immunoglobulin conjugated to Alexa Fluor™ 488. Liquid in Phosphate buffer with 15mg/mL BSA as a stabilizer and 0.1% sodium azide.
Gene Accession No. NM_001039538.1; NM_002374.3; NM_031845.2; NM_031847.2
Gene Symbols MAP2, MAP2B, MAP-2, MAP2C, DKFZp686I2148, MAP2A
Host Species Mouse
Immunogen Bovine brain microtubule protein.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Chicken, Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.