Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2 Mouse anti-Human, Phycoerythrin, Clone: AP20, MilliporeSigma™

Catalog No. FCMAB318PE Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB318PE 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB318PE Supplier MilliporeSigma Supplier No. FCMAB318PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Milli-Mark™ Anti-MAP2-PE Antibody, clone AP20 is an antibody against MAP2 for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP2
Applications Flow Cytometry
Classification Monoclonal
Clone AP20
Conjugate PE
Formulation Purified mouse monoclonal IgG1 conjugated to PE in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_002365
Gene Symbols MAP2
Host Species Mouse
Immunogen Recombinant Bovine brain microtubule protein.
Purification Method Protein A Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to 6 months. Protect from light.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.