Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2B Mouse, Alexa Fluor 647, Clone: 18/MAP2B, BD

Catalog No. BDB560382 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560382 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560382 Supplier BD Biosciences Supplier No. 560382
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Microtubule-associated proteins (MAPs) play a crucial role in the development and morphology of neurons. MAP2, specifically localized in dendrites, has four known isoforms that are produced by alternative splicing of the transcript and are expressed at various stages of neuronal development. MAP2B is a 280kDa protein that is expressed throughout brain development. It contains functional domains that interact with the regulatory subunit of the cAMP-dependent kinase II and microtubules. Experimental monitoring of its presence, along with GFAP and nestin, may be utilized to quantify neuronal development.

Host Species: Mouse
Clone: 18
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human MAP2B aa. 19-219

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Intracellular Staining, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP2B
Applications Flow Cytometry
Classification Monoclonal
Clone 18/MAP2B
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human MAP2B aa. 19-219
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.