Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mitogen-activated protein kinase kinase kinase 13, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016965 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-965 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-965 Supplier Abnova Corporation Supplier No. H00009175B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human MAP3K13 protein.

The protein encoded by this gene is a member of serine/threonine protein kinase family. This kinase contains a dual leucine-zipper motif, and has been shown to form dimers/oligomers through its leucine-zipper motif. This kinase can phosphorylate and activate MAPK8/JNK, MAP2K7/MKK7, which suggests a role in the JNK signaling pathway. [provided by RefSeq

Sequence: MANFQEHLSCSSSPHLPFSESKTFNGLQDELTAMGNHPSPKLLEDQQEKGMVRTELIESVHSPVTTTVLTSVSEDSRDQFENSVLQLREHDESETAVSQGNSNTVDGESTSGTEDIKIQFSRSGSGSGGFLEGLFGCLRPVWNIIGKAYSTDYKLQQQDTWEVPFEEISELQWLGSGAQGAVFLGKFRAEEVAIKKVREQNETDIKHLRKLKHPNIIAFKGVCTQAPCYCIIMEYCAHGQLYEVLRAGRKITPRLLVDWSTGIASGMNYLHLHKIIHRDLKSPNVLVTHTDAVKISDFGTSKELSDKSTKMSFAGTVAWMAPEVIRNEPVSEKVDIWSFGVVLWELLTGEIPYKDVDSSAIIWGVGSNSLHLPVPSTCPDGFKILMKQTWQSKPRNRPSFRQTLMHLDIASADVLATPQETYFKSQAEWREEVKKHFEKIKSEGTCIHRLDEELIRRRREELRHALDIREHYERKLERANNLYMELSAIMLQLEMREKELIKREQAVEKKYPGTYKRHPVRPIIHPNAMEKLMKRKGVPHKSGMQTKRPDLLRSEGIPTTEVAPTASPLSGSPKMSTSSSKSRYRSKPRHRRGNSRGSHSDFAAILKNQPAQENSPHPTYLHQAQSQYPSLHHHNSLQQQYQQPPPAMSQSHHPRLNMHGQDIATCANNLRYFGPAAALRSPLSNHAQRQLPGSSPDLISTAMAADCWRSSEPDKGQAGPWGCCQADAYDPCLQCRPEQYGSLDIPSAEPVGRSPDLSKSPAHNPLLENAQSSEKTEENEFSGCRSESSLGTSHLGTPPALPRKTRPLQKSGDDSSEEEEGEVDSEVEFPRRQRPHRCISSCQSYSTFSSENFSVSDGEEGNTSDHSNSPDELADKLEDRLAEKLDDLLSQTPEIPIDISSHSDGLSDKECAVRRVKTQMSLGKLCVEERGYENPMQFEESDCDSSDGECSDATVRTNKHYSSATW

Specifications

Antigen mitogen-activated protein kinase kinase kinase 13
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human MAP3K13 protein.
Formulation PBS with no preservative; pH 7.4
Gene MAP3K13
Gene Accession No. NM_004721.3
Gene Alias LZK/MGC133196
Gene Symbols MAP3K13
Host Species Mouse
Immunogen MAP3K13 (NP_004712.1, 1 a.a. ∼ 966 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Kinases
Primary or Secondary Primary
Gene ID (Entrez) 9175
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.