Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP3K5 Mouse anti-Human, Clone: 2E4, MilliporeSigma™

Catalog No. MABC632MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC632MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC632MI Supplier MilliporeSigma Supplier No. MABC632
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MAP3K5 Antibody, clone 2E4 is a highly specific mouse monoclonal antibody, that targets MAP3K5 and has been tested in western blotting, immunohistochemistry, immunocytochemistry, Flow Cytometry and ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP3K5
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2E4
Conjugate Unconjugated
Formulation Mouse monoclonal IgG2b ascitic fluid containing up to 0.1% sodium azide.
Gene Accession No. NP_005914
Gene Symbols MAP3K5, ASK1, MAPKKK5, MEKK5
Host Species Mouse
Immunogen Purified recombinant fragment of human MAP3K5 expressed in E. coli.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.